3H-Imidazo[4,5-b]pyridine-5-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A149516-10G
A149516-250MG
A149516-1G
A149516-25G
A149516-5G
103804-054EA
25
USD
103804-054
103842-170
76823-788
76823-790
77317-672
77317-674
77317-676
3H-Imidazo[4,5-b]pyridine-5-carboxylic acid 98%
3H-Imidazo[4,5-b]pyridine-5-carboxylic acid
Precaución: for Research Use Only
Formula:
C₇H₅N₃O₂ MW: 163.134 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09863858 CAS Number: 1019108-05-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...