3-Furoic acid 98%

Supplier: Ambeed
Warning

Synonyms: Furan-3-carboxylic acid, 3-Furancarboxylic acid

A147073-100G A147073-10G A147073-500G A147073-1G A147073-5G A147073-25G
103803-732EA 9 USD
103803-732 103841-828 76972-878 76972-880 76972-882 77299-562 77299-564 77317-186
3-Furoic acid 98%
3-Furoic acid

Precaución: for Research Use Only

Formula: C₅H₄O₃
MW: 112.08 g/mol
Boiling Pt: 229.6 °C (760 mm Hg)
Melting Pt: 120…122 °C
Flash Pt: 92.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00005350
CAS Number: 488-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR