3-Furanmethanol 97%

Supplier: Ambeed
Danger

A682066-1G A682066-25G A682066-5G A682066-10G
103840-540EA 39 USD
103840-540 76844-738 76844-740 76844-742 77280-724
3-Furanmethanol 97%
3-Furanmethanol

Precaución: for Research Use Only

Formula: C₅H₆O₂
Boiling Pt: 180.3 °C (760 mm Hg)
Flash Pt: 38 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005352
CAS Number: 4412-91-3
UN: 1987
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR