Furan-3-boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Furanboronic acid, 3-Furanylboronic acid

A185779-100G A185779-1G A185779-25G
103815-882EA 7 USD
103815-882 103846-452 76872-880 77084-086 77301-992
Furan-3-boronic acid 97%
Furan-3-boronic acid

Precaución: for Research Use Only

Formula: C₄H₅BO₃
MW: 111.89 g/mol
Boiling Pt: 247.7 °C (760 mm Hg)
Melting Pt: 139…144 °C
Flash Pt: 103.6 °C
Storage Temperature: Ambient
MDL Number: MFCD01319007
CAS Number: 55552-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR