3-Formylbenzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: Isophthalaldehydic acid, 3-Carboxybenzaldehyde

A104474-10G A104474-25G A104474-1G A104474-5G
103796-716EA 6 USD
103796-716 103822-430 77270-418 77270-420 77309-296 77309-298
3-Formylbenzoic acid 98%
3-Formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆O₃
MW: 150.13 g/mol
Boiling Pt: 331.7 °C (760 mm Hg)
Melting Pt: 172…176 °C
Flash Pt: 168.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00039575
CAS Number: 619-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR