3-Formyl-5-methylbenzonitrile 97%

Supplier: Ambeed
Warning

A189735-5G A189735-100MG A189735-1G A189735-250MG
103846-814EA 512 USD
103846-814 77084-586 77191-946 77191-948 77191-950
3-Formyl-5-methylbenzonitrile 97%
3-Formyl-5-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₉H₇NO
MW: 145.15 g/mol
Boiling Pt: 240.0±20.0 °C (760 mm Hg)
Flash Pt: 99 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18382518
CAS Number: 27633-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR