3-Formyl-4-methoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Formyl-4-methoxybenzeneboronic acid

A230769-1G A230769-25G A230769-10G
103810-728EA 9 USD
103810-728 103850-472 77089-344 77089-346 77249-570
3-Formyl-4-methoxyphenylboronic acid 98%
3-Formyl-4-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BO₄
MW: 179.97 g/mol
Melting Pt: 214…217 °C
Flash Pt: 192.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093661
CAS Number: 121124-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR