3-Formyl-2-methoxyphenylboronic acid 95%

Supplier: Ambeed
Warning

A248464-250MG A248464-1G A248464-5G
103811-824EA 19 USD
103811-824 103851-658 77031-448 77091-002 77091-004
3-Formyl-2-methoxyphenylboronic acid 95%
3-Formyl-2-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BO₄
MW: 179.9700 g/mol
Boiling Pt: 407.7 °C (760 mm Hg)
Melting Pt: 122…125 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08689486
CAS Number: 480424-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR