3-Fluoropyrrolidine hydrochloride 97%

Supplier: Ambeed
Warning

A278152-100G A278152-1G A278152-25G A278152-5G A278152-250MG
103813-398EA 13 USD
103813-398 103853-336 77032-120 77093-246 77093-248 77093-250 77668-214
3-Fluoropyrrolidine hydrochloride 97%
3-Fluoropyrrolidine hydrochloride

Precaución: for Research Use Only

Formula: C4H9ClFN
Storage Temperature: Refrigerator
MDL Number: MFCD04038329
CAS Number: 169750-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR