3-Fluoropyridine-2-carbonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-2-pyridinecarbonitrile, 2-Cyano-3-fluoropyridine, 3-Fluoropicolinonitrile

A329706-25G A329706-5G A329706-100G A329706-10G A329706-500G A329706-1G
103825-536EA 9 USD
103825-536 103836-492 76839-462 76839-464 77141-930 77141-932 77141-934 77306-402
3-Fluoropyridine-2-carbonitrile 97%
3-Fluoropyridine-2-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₃FN₂
MW: 122.1 g/mol
Melting Pt: 27…32 °C
Flash Pt: 104.4 °C
Storage Temperature: Ambient
MDL Number: MFCD06797501
CAS Number: 97509-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR