3-Fluorophenol 98%

Supplier: Ambeed
Danger

A185840-25G A185840-5G A185840-100G
103846-460EA 14 USD
103846-460 77084-098 77084-100 77593-194
3-Fluorophenol 98%
3-Fluorophenol

Precaución: for Research Use Only

Formula: C₆H₅FO
MW: 112.1 g/mol
Boiling Pt: 178 °C (760 mm Hg)
Melting Pt: 8…12 °C
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00002254
CAS Number: 372-20-3
UN: 2922
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR