3-Fluoroisonicotinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoropyridine-4-carboxylic acid

A171118-1G A171118-25G A171118-5G A171118-100G A171118-10G A171118-250MG
103806-592EA 6 USD
103806-592 103844-866 77081-992 77081-994 77081-996 77096-522 77096-524 77096-526
3-Fluoroisonicotinic acid 98%
3-Fluoroisonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₄FNO₂
MW: 141.1 g/mol
Boiling Pt: 360 °C (760 mm Hg)
Melting Pt: 251…254 °C
Flash Pt: 171.5 °C
Storage Temperature: Ambient
MDL Number: MFCD03789251
CAS Number: 393-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR