3-Fluoropyrocatechol 98%

Supplier: Ambeed
Danger

Synonyms: 3-Fluorocatechol, 3-Fluorobenzene-1,2-diol, 3-Fluoro-1,2-dihydroxybenzene

A353008-1G A353008-25G A353008-5G A353008-100G A353008-10G A353008-250MG
103826-084EA 12 USD
103826-084 103837-086 76840-196 76840-198 76840-200 77290-684 77290-686 77290-688
3-Fluoropyrocatechol 98%
3-Fluoropyrocatechol

Precaución: for Research Use Only

Formula: C₆H₅FO₂
MW: 128.1 g/mol
Boiling Pt: 218.4 °C (760 mm Hg)
Melting Pt: 71…73 °C
Flash Pt: 85.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00042582
CAS Number: 363-52-0
UN: 1759
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR