3-Fluorobenzyl alcohol 96%

Supplier: Ambeed
Warning

Synonyms: (3-Fluorophenyl)methanol

A206516-5G A206516-100G A206516-10G A206516-25G
103848-386EA 6 USD
103848-386 77086-648 77097-564 77097-566 77302-988
3-Fluorobenzyl alcohol 96%
3-Fluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₇FO
Boiling Pt: 204.5 °C (760 mm Hg)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00004631
CAS Number: 456-47-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR