3-Fluorobenzoic acid 97%

Supplier: Ambeed
Warning

A160641-100G A160641-10G A160641-25G A160641-500G A160641-1000G
103843-632EA 5 USD
103843-632 76768-544 76768-546 76768-548 76768-550 77598-114
3-Fluorobenzoic acid 97%
3-Fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅FO₂
Melting Pt: 122…124 °C
Flash Pt: 105.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00002489
CAS Number: 455-38-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR