3-Fluorobenzamide 99%

Supplier: Ambeed
Warning

A589418-25G A589418-5G A589418-100G
103840-066EA 18 USD
103840-066 76844-214 76844-216 77280-206
3-Fluorobenzamide 99%
3-Fluorobenzamide

Precaución: for Research Use Only

Formula: C₇H₆FNO
Boiling Pt: 238.4 °C (760 mm Hg)
Melting Pt: 129…132 °C at N/A mmHg
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00007983
CAS Number: 455-37-8
ADR: N/A,N/A

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR