α,α,α,5-Tetrafluoro-m-toluic acid 98%
Supplier: Ambeed
|
Warning
|
A139085-100G
A139085-10G
A139085-1G
A139085-25G
A139085-5G
103802-692EA
9
USD
103802-692
76767-692
76767-694
77315-766
77315-768
77315-770
α,α,α,5-Tetrafluoro-m-toluic acid 98%
α,α,α,5-Tetrafluoro-m-toluic acid
Precaución: for Research Use Only
Formula:
C₈H₄F₄O₂ Boiling Pt: 239.6 °C (760 mm Hg) Melting Pt: 103…105 °C Flash Pt: 98.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00061293 CAS Number: 161622-05-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...