3-Fluoro-5-nitrobenzaldehyde 97%

Supplier: Ambeed
Warning

A103370-10G A103370-250MG A103370-25G A103370-1G A103370-5G
103796-538EA 24 USD
103796-538 103822-242 76780-632 76780-634 76780-636 77283-084 77283-086
3-Fluoro-5-nitrobenzaldehyde 97%
3-Fluoro-5-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄FNO₃
Boiling Pt: 265 °C (760 mm Hg)
Flash Pt: 114 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11111000
CAS Number: 108159-96-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR