3-Fluoro-5-nitro-1-trifluoromethylbenzene 98%

Supplier: Ambeed
Warning

A198580-10G A198580-1G A198580-25G A198580-5G
103817-034EA 12 USD
103817-034 103847-662 77274-732 77302-592 77302-594 77302-596
3-Fluoro-5-nitro-1-trifluoromethylbenzene 98%
3-Fluoro-5-nitro-1-trifluoromethylbenzene

Precaución: for Research Use Only

Formula: C7H3F4NO2
MW: 209.0978 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07779168
CAS Number: 454-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR