5-Fluoro-m-toluonitrile 97%

Supplier: Ambeed
Warning

A786468-10G A786468-250MG A786468-1G A786468-25G
103840-998EA 14 USD
103840-998 76776-474 76776-476 76845-250 77308-328
5-Fluoro-m-toluonitrile 97%
5-Fluoro-m-toluonitrile

Precaución: for Research Use Only

Formula: C₈H₆FN
MW: 135.14 g/mol
Melting Pt: 53…56 °C
Storage Temperature: Ambient
MDL Number: MFCD03094312
CAS Number: 216976-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR