3-Fluoro-5-methylbenzaldehyde 97%

Supplier: Ambeed
Warning

A175979-10G A175979-1G A175979-25G A175979-5G
103807-106EA 20 USD
103807-106 103845-418 76690-352 77301-412 77301-414 77301-416
3-Fluoro-5-methylbenzaldehyde 97%
3-Fluoro-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇FO
Boiling Pt: 197.6 °C (760 mm Hg)
Flash Pt: 79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094315
CAS Number: 189628-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR