5-Fluoro-m-toluidine 97%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-5-methylaniline

A274943-1G A274943-25G
103813-234EA 6 USD
103813-234 103853-162 77093-000 77093-002
5-Fluoro-m-toluidine 97%
5-Fluoro-m-toluidine

Precaución: for Research Use Only

Formula: C₇H₈FN
MW: 125.14 g/mol
Boiling Pt: 207.4 °C (760 mm Hg)
Flash Pt: 89.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00077538
CAS Number: 52215-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR