3-Fluoro-5-methoxybenzoic acid 98%

Supplier: Ambeed
Warning

A247191-10G A247191-1G A247191-25G A247191-5G
103811-754EA 6 USD
103811-754 103851-586 76976-602 77090-900 77090-902 77090-904
3-Fluoro-5-methoxybenzoic acid 98%
3-Fluoro-5-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇FO₃
MW: 170.1400 g/mol
Melting Pt: 130…133 °C
Storage Temperature: Ambient
MDL Number: MFCD08235092
CAS Number: 176548-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR