5-Fluoro-m-anisaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-5-methoxybenzaldehyde, 5-Fluoro-3-methoxybenzaldehyde, 3-Fluoro-5-anisaldehyde

A136939-100G A136939-10G A136939-1G A136939-25G A136939-5G
103802-378EA 10 USD
103802-378 76688-834 76688-836 77315-304 77315-306 77315-308
5-Fluoro-m-anisaldehyde 98%
5-Fluoro-m-anisaldehyde

Precaución: for Research Use Only

Formula: C₈H₇FO₂
MW: 154.14 g/mol
Boiling Pt: 216 °C (760 mm Hg)
Flash Pt: 82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08741376
CAS Number: 699016-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR