3-Fluoro-5-methoxyaniline 98%

Supplier: Ambeed
Warning

Synonyms: 5-Fluoro-m-anisidine, 3-Amino-5-fluoroanisole

A144210-1G A144210-10G
103803-380EA 12 USD
103803-380 103841-456 77299-370 77667-078
3-Fluoro-5-methoxyaniline 98%
3-Fluoro-5-methoxyaniline

Precaución: for Research Use Only

Formula: C₇H₈FNO
MW: 141.143 g/mol
Flash Pt: 90.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077535
CAS Number: 2339-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR