3-Fluoro-5-iodopyridin-2-amine 95%

Supplier: Ambeed
Warning

A297963-1G A297963-5G A297963-100MG A297963-250MG A297963-25G
103814-166EA 17 USD
103814-166 103835-310 76837-998 76838-000 77166-916 77166-918 77305-734
3-Fluoro-5-iodopyridin-2-amine 95%
3-Fluoro-5-iodopyridin-2-amine

Precaución: for Research Use Only

Formula: C₅H₄FIN₂
Storage Temperature: Refrigerator
MDL Number: MFCD22628097
CAS Number: 1321612-85-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR