3-Fluoro-5-iodobenzonitrile 98%

Supplier: Ambeed
Warning

A170220-1G A170220-100MG A170220-250MG
103806-458EA 42 USD
103806-458 103844-728 77081-836 77883-316 77883-317
3-Fluoro-5-iodobenzonitrile 98%
3-Fluoro-5-iodobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃FIN
Boiling Pt: 251 °C (760 mm Hg)
Melting Pt: 29…30 °C
Flash Pt: 105 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04116468
CAS Number: 723294-75-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR