3-Fluoro-5-hydroxybenzoic acid 97%

Supplier: Ambeed
Warning

A434653-25G A434653-10G A434653-250MG A434653-1G A434653-100G
103827-582EA 12 USD
103827-582 103838-648 76842-366 76979-328 76979-330 77307-282 77594-354
3-Fluoro-5-hydroxybenzoic acid 97%
3-Fluoro-5-hydroxybenzoic acid

Precaución: For research use only

Formula: C₇H₅FO₃
Boiling Pt: 351.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD11846515
CAS Number: 860296-12-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR