3-Fluoro-5-formylbenzonitrile 98%

Supplier: Ambeed
Warning

A250649-100G A250649-250MG A250649-1G A250649-25G A250649-5G
103811-938EA 14 USD
103811-938 103851-776 76786-812 76786-814 77091-130 77091-132 77091-134
3-Fluoro-5-formylbenzonitrile 98%
3-Fluoro-5-formylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₄FNO
MW: 149.1200 g/mol
Boiling Pt: 205 °C (760 mm Hg)
Flash Pt: 77 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04974123
CAS Number: 1003708-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR