3-Fluoro-5-(trifluoromethyl)phenylacetonitrile 98%
Supplier: Ambeed
|
Warning
|
A163448-10G
A163448-1G
A163448-25G
103805-728EA
29
USD
103805-728
103843-988
77096-226
77300-660
77320-086
3-Fluoro-5-(trifluoromethyl)phenylacetonitrile 98%
3-Fluoro-5-(trifluoromethyl)phenylacetonitrile
Precaución: for Research Use Only
Formula:
C₉H₅F₄N MW: 203.1400 g/mol Boiling Pt: 205.8 °C (760 mm Hg) Flash Pt: 89.6 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00061181 CAS Number: 239087-12-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...