3-Fluoro-5-isobutoxybenzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-5-(isobutoxy)benzeneboronicacid

A273756-1G A273756-5G
103813-160EA 14 USD
103813-160 103853-084 77305-148 77305-150
3-Fluoro-5-isobutoxybenzeneboronic acid 98%
3-Fluoro-5-isobutoxybenzeneboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₄BFO₃
MW: 212.03 g/mol
Boiling Pt: 334.9 °C (760 mm Hg)
Flash Pt: 156.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07363783
CAS Number: 850589-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR