3-Fluoro-4-nitrobenzonitrile 98%

Supplier: Ambeed
Warning

A113810-100G A113810-10G A113810-25G A113810-1G
103798-294EA 9 USD
103798-294 103824-138 76766-030 76766-032 76766-034 77310-956
3-Fluoro-4-nitrobenzonitrile 98%
3-Fluoro-4-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃FN₂O₂
Boiling Pt: 316.4 °C (760 mm Hg)
Melting Pt: 72…73 °C
Flash Pt: 145.2 °C
Storage Temperature: Ambient
MDL Number: MFCD04973417
CAS Number: 218632-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR