3-Fluoro-4-nitroaniline 98%

Supplier: Ambeed
Warning

A228187-5G A228187-10G
103850-278EA 17 USD
103850-278 77089-072 77098-156
3-Fluoro-4-nitroaniline 98%
3-Fluoro-4-nitroaniline

Precaución: for Research Use Only

Formula: C₆H₅FN₂O₂
MW: 156.12
Boiling Pt: 335.2 °C (760 mm Hg)
Melting Pt: 158…162 °C
Flash Pt: 156.6 °C
Storage Temperature: Ambient
MDL Number: MFCD03412239
CAS Number: 2369-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR