3-Fluoro-p-cresol 97%

Supplier: Ambeed
Warning

A139276-10G A139276-1G A139276-25G A139276-5G A139276-100G
103802-724EA 6 USD
103802-724 76767-696 77299-070 77315-808 77315-810 77876-968
3-Fluoro-p-cresol 97%
3-Fluoro-p-cresol

Precaución: for Research Use Only

Formula: C₇H₇FO
Boiling Pt: 196 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00143164
CAS Number: 452-78-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR