3-Fluoro-p-toluic acid 98%

Supplier: Ambeed
Warning

A139896-100G A139896-10G A139896-1G A139896-25G A139896-5G
103802-792EA 6 USD
103802-792 77272-454 77272-456 77315-894 77315-896 77315-898
3-Fluoro-p-toluic acid 98%
3-Fluoro-p-toluic acid

Precaución: for Research Use Only

Formula: C₈H₇FO₂
MW: 154.14 g/mol
Boiling Pt: 275.3 °C (760 mm Hg)
Melting Pt: 169…171 °C
Flash Pt: 120.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00002490
CAS Number: 350-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR