3-Fluoro-4-methylphenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-4-methylbenzeneboronic acid

A277234-1G A277234-25G A277234-5G A277234-10G
103813-342EA 9 USD
103813-342 103853-276 77093-164 77093-166 77093-168 77194-076
3-Fluoro-4-methylphenylboronic acid 97%
3-Fluoro-4-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BFO₂
Melting Pt: 232…237 °C
Flash Pt: 162.5 °C
Storage Temperature: Ambient
MDL Number: MFCD02683115
CAS Number: 168267-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR