(3-Fluoro-4-methoxyphenyl)acetonitrile 98%

Supplier: Ambeed
Danger

Synonyms: 3-Fluoro-4-methoxylphenylacetonitrile

A299316-1G A299316-5G A299316-100MG A299316-10G A299316-250MG
103814-222EA 8 USD
103814-222 103835-368 76838-060 76838-062 76977-694 76977-696 76977-698
(3-Fluoro-4-methoxyphenyl)acetonitrile 98%
(3-Fluoro-4-methoxyphenyl)acetonitrile

Precaución: For research use only

Formula: C₉H₈FNO
MW: 165.17 g/mol
Boiling Pt: 270.6 °C (760 mm Hg)
Melting Pt: 46…48 °C
Flash Pt: 117.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00040889
CAS Number: 404-90-0
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR