3-Fluoro-4-methoxybenzonitrile 96%

Supplier: Ambeed
Warning

A498996-100G A498996-10G A498996-1G A498996-250MG A498996-25G A498996-5G
103839-362EA 10 USD
103839-362 76774-474 76774-476 76774-478 76774-480 76843-330 76843-332
3-Fluoro-4-methoxybenzonitrile 96%
3-Fluoro-4-methoxybenzonitrile

Precaución: for Research Use Only

Formula: C₈H₆FNO
MW: 151.14 g/mol
Boiling Pt: 254.3 °C (760 mm Hg)
Melting Pt: 98…100 °C
Flash Pt: 107.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00215835
CAS Number: 331-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR