3-Fluoro-p-anisaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-4-methoxybenzaldehyde, 3-Fluoro-4-anisaldehyde

A127861-100G A127861-1G A127861-25G A127861-10G
103801-030EA 6 USD
103801-030 76688-518 77298-246 77313-304 77876-916
3-Fluoro-p-anisaldehyde 97%
3-Fluoro-p-anisaldehyde

Precaución: for Research Use Only

Formula: C₈H₇FO₂
Boiling Pt: 247.1 °C (760 mm Hg)
Melting Pt: 34…35 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003349
CAS Number: 351-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR