3-Fluoro-4-iodopyridine 97%
Supplier: Ambeed
|
Warning
|
A476901-1G
A476901-25G
A476901-5G
A476901-100G
A476901-10G
103828-088EA
9
USD
103828-088
103839-158
76843-088
76843-090
76843-092
77016-992
77016-994
3-Fluoro-4-iodopyridine 97%
3-Fluoro-4-iodopyridine
Precaución: for Research Use Only
Formula:
C₅H₃FIN Boiling Pt: 435.8 °C (760 mm Hg) Flash Pt: 76.6 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03092927 CAS Number: 22282-75-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...