3-Fluoro-4-iodobenzonitrile 98%
Supplier: Ambeed
|
Warning
|
A115733-100G
A115733-25G
A115733-1G
A115733-5G
103798-608EA
11
USD
103798-608
103824-468
76766-152
76766-154
77311-286
77311-288
3-Fluoro-4-iodobenzonitrile 98%
3-Fluoro-4-iodobenzonitrile
Precaución: for Research Use Only
Formula:
C₇H₃FIN MW: 247.01 g/mol Melting Pt: 113…117 °C Storage Temperature: Ambient |
MDL Number:
MFCD07782076 CAS Number: 887266-99-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...