3-Fluoro-4-hydroxy-5-methoxybenzaldehyde 97%

Supplier: Ambeed
Warning

A136878-250MG A136878-5G
103802-366EA 71 USD
103802-366 77234-750 77315-284
3-Fluoro-4-hydroxy-5-methoxybenzaldehyde 97%
3-Fluoro-4-hydroxy-5-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇FO₃
Melting Pt: 114…118 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02683560
CAS Number: 79418-78-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR