3-Fluoro-4-cyanopyridine 98%

Supplier: Ambeed
Warning

A141797-10G A141797-250MG A141797-1G A141797-25G A141797-5G
103803-060EA 37 USD
103803-060 76767-804 76767-806 77316-250 77316-252 77316-254
3-Fluoro-4-cyanopyridine 98%
3-Fluoro-4-cyanopyridine

Precaución: for Research Use Only

Formula: C6H3FN2
Boiling Pt: 178.8 °C (760 mm Hg)
Flash Pt: 61.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04112524
CAS Number: 113770-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR