α,α,α,3-Tetrafluoro-p-toluic acid 98%
Supplier: Ambeed
A112121-100G
A112121-10G
A112121-25G
A112121-1G
103797-956EA
7
USD
103797-956
103823-790
77189-122
77189-124
77189-126
77310-598
α,α,α,3-Tetrafluoro-p-toluic acid 98%
α,α,α,3-Tetrafluoro-p-toluic acid
Precaución: for Research Use Only
Formula:
C₈H₄F₄O₂ |
MDL Number:
MFCD00236279 CAS Number: 115754-21-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...