3-Fluoro-4-(trifluoromethoxy)phenol 98%
Supplier: Ambeed
|
Warning
|
A103395-10G
A103395-25G
A103395-1G
A103395-5G
A103395-250MG
103796-544EA
23
USD
103796-544
103822-248
77270-348
77270-350
77309-114
77309-116
77747-769
3-Fluoro-4-(trifluoromethoxy)phenol 98%
3-Fluoro-4-(trifluoromethoxy)phenol
Precaución: for Research Use Only
Formula:
C₇H₄F₄O₂ Boiling Pt: 196.9 °C (760 mm Hg) Flash Pt: 99.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD06660342 CAS Number: 177596-38-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...