3-Fluoro-4'-(methylsulfonyl)-[1,1'-biphenyl]-4-ol 95%

Supplier: Ambeed
Warning

A481443-1G A481443-100MG A481443-250MG
103828-128EA 128 USD
103828-128 76843-146 76877-842 76877-844
3-Fluoro-4'-(methylsulfonyl)-[1,1'-biphenyl]-4-ol 95%
3-Fluoro-4'-(methylsulfonyl)-[1,1'-biphenyl]-4-ol

Precaución: for Research Use Only

Formula: C₁₃H₁₁FO₃S
MW: 266.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18313997
CAS Number: 1032825-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR