4-Amino-2-fluorophenylboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-2-fluorobenzeneboronic acid pinacol ester, 3-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

A140072-100MG A140072-10G A140072-250MG A140072-1G A140072-25G A140072-5G
103802-828EA 21 USD
103802-828 77285-702 77285-704 77285-706 77299-116 77299-118 77299-120
4-Amino-2-fluorophenylboronic acid pinacol ester 97%
4-Amino-2-fluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₇BFNO₂
Flash Pt: 161.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09951877
CAS Number: 819057-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR