3-Fluoro-2-nitrotoluene 98%

Supplier: Ambeed
Warning

A122102-100G A122102-10G A122102-25G A122102-500G A122102-1G A122102-5G
103800-176EA 6 USD
103800-176 76781-816 76781-818 76781-820 76781-822 77312-306 77312-308
3-Fluoro-2-nitrotoluene 98%
3-Fluoro-2-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₆FNO₂
Melting Pt: 17…18 °C
Flash Pt: 90.5 °C
Storage Temperature: Ambient
MDL Number: MFCD08458093
CAS Number: 3013-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR