3-Fluoro-2-nitropyridine 98%
Supplier: Ambeed
|
Warning
|
A214434-100G
A214434-100MG
A214434-10G
A214434-250MG
A214434-5G
A214434-1G
A214434-25G
103818-402EA
11
USD
103818-402
103849-108
76975-632
76975-634
76975-636
76975-638
77087-630
77303-344
77303-346
3-Fluoro-2-nitropyridine 98%
3-Fluoro-2-nitropyridine
Precaución: for Research Use Only
Formula:
C₅H₃FN₂O₂ Boiling Pt: 255.8 °C (760 mm Hg) Melting Pt: 35 °C Flash Pt: 108.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04114127 CAS Number: 54231-35-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...