3-Fluoro-2-nitrobenzonitrile 97%

Supplier: Ambeed
Warning

A127720-100MG A127720-1G A127720-250MG A127720-5G
76971-930EA 13.8 USD
76971-930 76971-932 76971-934 77313-262
3-Fluoro-2-nitrobenzonitrile 97%
3-Fluoro-2-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃FN₂O₂
MW: 166.1100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09864664
CAS Number: 1000339-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR